Learn More
Manutan Shelving starter bay, chrome wire, 1880mm x 1220mm x 355mm
Description
A versatile shelving option where light and air distribution are important. Adjustable, simple to assemble and easy to clean. Suitable for use in food and medical areas where a high standard of hygiene must be maintained. Robust construction with shelf capacity of up to 200kg. Four adjustable shelves to suit specific storage requirements. Adjustable feet for uneven ground. Self assembly (no nuts and bolts required).
- Number of shelves : 4
- Shelf capacity UDL: 175kg
- Dimensions (H x W x D): 1880mm x 1220mm x 355mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.