Learn More
Manutan Shelving, Rapid 1 extra heavy duty, blue/orange, four shelves, 1980mm x 2134mm x 915mm
Product Code. 12832148
Encompass_Preferred
Product Code. 12832148
Brand: Manutan A361070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Industrial strength frame with shelving loads of 500kg-640kg. Simple boltless assembly - frames and beams simply clip together and only require a rubber mallet. Shelves adjustable by 38mm increments, even when built. High density chipboard shelves with max shelf capacity of up to 640kg. 2mm European structural quality steel. Hardwearing powder coated finish. Load capacity TUV certified - test to establish safe working loads.
  • Shelf capacity UDL: 580kg
  • Dimensions (W x D): 2134mm x 915mm
  • Four shelves