missing translation for 'onlineSavingsMsg'
Learn More

Justrite Shelf for EN Cabinet 226-EN 2903, Solid

Product Code. 17742388
Click to view available options
Color:
Light Gray
Stainless Steel
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17742388

Brand: Justrite JCBS120IS0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Additional shelf accessory for EN Cabinets 226-EN. Choice of installed or pull out shelves in stainless steel or epoxy coated finishings.

  • Choice of stainless steel or epoxy coated shelves
  • Choice of installed or pull out shelves

Specifications

For Use With (Equipment) EN Cabinet 226-EN 2903
Length (Metric) 109 cm
Shelf Type Pull Out
Width (Metric) 42 cm
Color Stainless Steel
Material Steel
Product Type Shelf
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.