missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Sheep IL-5 ELISA Development Kit

Product Code. 18364324 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit (for 6 Plates)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364324 1 Kit (for 6 Plates) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364324 Supplier Novus Biologicals™ Supplier No. NBP318277

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 4-1000 pg/ml
Product Type ELISA Development Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
Specificity The kit contains a matched pair of monoclonal antibodies (mAbs) specific for native and recombinant bovine IL-5.
For Use With (Application) ELISA
Gene ID (Entrez) 3567
Gene Symbol IL5
Quantity 1 Kit (for 6 Plates)
Research Discipline Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I
Target Sheep
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.