Learn More
VAV2 Sheep anti-Human, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against partial recombinant VAV2.
Brand: Abnova PAB9710.250ug
Description
VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| VAV2 | |
| Polyclonal | |
| Sheep polyclonal antibody raised against partial recombinant VAV2. | |
| In PBS (0.08% sodium azide) | |
| VAV2 | |
| Recombinant His fusion protein corresponding to amino acids 287-578 of human VAV2. | |
| RUO | |
| 7410 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation | |
| Unconjugated | |
| Western Blot (1:5000) The optimal working dilution should be determined by the end user. | |
| VAV2 | |
| Sheep | |
| 250 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only