missing translation for 'onlineSavingsMsg'
Learn More

VAV2 Sheep anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16051858
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16051858 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16051858 Supplier Abnova Supplier No. PAB9711.250ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep polyclonal antibody raised against partial recombinant VAV2.

VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen VAV2
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against partial recombinant VAV2.
Dilution Western Blot (1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene VAV2
Gene Symbols VAV2
Host Species Sheep
Immunogen Recombinant His fusion protein corresponding to amino acids 578-878 of human VAV2.
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7410
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.