missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINC1, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against native Serpinc1.
Brand: Abnova PAB0688.1mg
This item is not returnable.
View return policy
Specifications
| SERPINC1 | |
| Polyclonal | |
| Sheep polyclonal antibody raised against native Serpinc1. | |
| In PBS (50% glycerol) | |
| AI114908/ATIII/At-3/At3 | |
| Sheep | |
| 1 mg | |
| Primary | |
| This antibody recognizes mouse antithrombin. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (25 ug/mL) The optimal working dilution should be determined by the end user. | |
| Serpinc1 | |
| Serpinc1 | |
| Native purified Mouse Serpinc1. | |
| RUO | |
| 11905 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction