missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGF2-alpha, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against PGF2-alpha.
Brand: Abnova PAB13625.100uL
This item is not returnable.
View return policy
Specifications
| PGF2-alpha | |
| Polyclonal | |
| Sheep polyclonal antibody raised against PGF2-alpha. | |
| In antiserum | |
| PGF2-alpha. | |
| RUO | |
| PGF2a 100% 6-keto PGF1a 0.9% Thromboxane B2 16% 13, 14-dihydro-15-keto PGF2a 0.06% 13, 14-dihydro-6.15diketo PGF2a <0.01% PGD2 0.2% PGE1 0.1% PGE2 0.3% 6-keto PGE1 0.03% PGF1a 21% Arachidonic acid <0.001% | |
| Liquid |
| Radioimmune Assays (RIA) | |
| Unconjugated | |
| Radioimmunoassay (1:10000) The optimal working dilution should be determined by the end user. | |
| Sheep | |
| 100 μL | |
| Primary | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction