Learn More
Sheep anti-Mouse IgG2a Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69579
Product Code. 18602817
  • £296.00 / 1mL
Description
IgG2a Polyclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Immunoelectrophoresis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG2a
Polyclonal
Unconjugated
Sheep
Whole Antiserum
RUO
Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunoelectrophoresis
LYOPH
Whole antiserum
Highly purified normal homogenous IgG2a(1b) isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
Secondary
Mouse
Lyophilized