missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
pIgR Polyclonal antibody specifically detects pIgR in Human samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Immunoblot, Dot Blot
Specifications
Specifications
| Antigen | pIgR |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Immunoblot, Dot Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry, Immunoprecipitation 1:10 - 1:500, Immunoblotting, Dot Blot |
| Formulation | Lyophilized from PBS (pH 7.2) |
| Gene Alias | FLJ22667, hepatocellular carcinoma associated protein TB6, Hepatocellular carcinoma-associated protein TB6, MGC125361, MGC125362, PIgR, Poly-Ig receptor, polymeric immunoglobulin receptor |
| Host Species | Sheep |
| Immunogen | Purified free human secretory component isolated from pooled milk is used for immunization. Freund's complete adjuvant is used in the first step of the immunization procedure. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?