missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Human IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB120-6870
189.55 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| HRP | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2.0 mg/mL | |
| Western Blot 1:1000 - 1:10000, ELISA 1:150000 - 1:250000, Immunohistochemistry 1:500 - 1:2500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Human IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Prepared from monospecific antiserum by immunoaffinity chromatography using Human IgG coupled to agarose. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Sheep Serum, Human IgG and Human Serum. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction