missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Human IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP1-72572
205.70 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot, Electron Microscopy.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot, Immunoassay | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200 - 1:1000, Dot Blot, Electron Microscopy | |
| Human IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography using Human IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Alkaline Phosphatase (calf intestine), anti-Sheep Serum, Human IgG and Human Serum. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction