Learn More
FOS Sheep anti-Hamster, Human, Rabbit, Rat, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against synthetic peptide of FOS.
Brand: Abnova PAB0751.500ug
Description
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death. [provided by RefSeq
Sequence: MFSGFNADYEASSSRCSpecifications
| FOS | |
| Polyclonal | |
| Sheep polyclonal antibody raised against synthetic peptide of FOS. | |
| Lyophilized from PBS | |
| P01100;Q4FDN1;P01101;Q56TL8;Q5XQC6 | |
| FOS | |
| A synthetic peptide (conjugated diphtheria toxoid) corresponding to amino acids 2-17 of human FOS. | |
| 500 μg | |
| Primary | |
| This antiserum shows a high level of specificity for cFOS confirmed by immunohostochemistry. | |
| Store at -20°C. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (10 ug/mL) Western Blot (10 ug/mL) The optimal dilution should be determined by the end user. | |
| FOS | |
| AP-1/C-FOS | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2353 | |
| Hamster, Human, Rabbit, Rat | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only