Learn More
DBH Sheep anti-Human, Primates, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against synthetic peptide of DBH.
Brand: Abnova PAB9693.100uL
Description
The protein encoded by this gene is an oxidoreductase belonging to the copper type II, ascorbate-dependent monooxygenase family. It is present in the synaptic vesicles of postganglionic sympathetic neurons and converts dopamine to norepinephrine. It exists in both soluble and membrane-bound forms, depending on the absence or presence, respectively, of a signal peptide. [provided by RefSeq
Specifications
| DBH | |
| Polyclonal | |
| Sheep polyclonal antibody raised against synthetic peptide of DBH. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| P09172 | |
| DBH | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus human DBH. | |
| 100 μL | |
| Primary | |
| Human, Primate | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| DBH | |
| DBM | |
| Sheep | |
| Affinity purification | |
| RUO | |
| 1621 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only