missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dab1 Sheep anti-Mouse, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against partial recombinant Dab1.
Brand: Abnova PAB9857.250ug
This item is not returnable.
View return policy
Specifications
| Dab1 | |
| Polyclonal | |
| Sheep polyclonal antibody raised against partial recombinant Dab1. | |
| In PBS (0.08% sodium azide) | |
| AI956902/C630028C02Rik/scm/scr/scrambler/yot | |
| Sheep | |
| 250 μg | |
| Primary | |
| Mouse | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1-5 ug/mL) Immunoprecipitation (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Dab1 | |
| Dab1 | |
| Recombinant GST fusion protein corresponding to amino acids 1-179 of mouse Dab1. | |
| RUO | |
| 13131 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction