missing translation for 'onlineSavingsMsg'
Learn More

cGMP, Sheep, Polyclonal Antibody, Abnova™

Product Code. 16042637
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16042637 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16042637 Supplier Abnova Supplier No. PAB17110.200uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep polyclonal antibody raised against cGMP.

Specifications

Antigen cGMP
Applications ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against cGMP.
Dilution Radioimmunoassay (1:6400) The optimal working dilution should be determined by the end user.
Formulation In serum
Host Species Sheep
Immunogen Succinylated cGMP conjugated to bovine serum albumin.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts specifically with cyclic GMP.
Target Species Bovine
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.