missing translation for 'onlineSavingsMsg'
Learn More

Bovine Serum Albumin, BSA Sheep, Polyclonal Antibody, Abnova™

Product Code. 16068976
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16068976 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16068976 Supplier Abnova Supplier No. PAB0903.200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep polyclonal antibody raised against Bovine Serum Albumin, BSA.

Specifications

Antigen Bovine Serum Albumin, BSA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against Bovine Serum Albumin, BSA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.09% sodium azide)
Host Species Sheep
Immunogen Bovine Serum Albumin, BSA.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.