missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgG2 Secondary Antibody, DyLight 594, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG2 Polyclonal antibody specifically detects IgG2 in Bovine samples. It is validated for ELISA, Immunodiffusion
Specifications
Specifications
| Antigen | IgG2 |
| Applications | ELISA, Immunodiffusion |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker) |
| Host Species | Sheep |
| Immunogen | Purified bovine IgG2 |
| Purification Method | Affinity purified |
| Quantity | 1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?