missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgA Secondary Antibody, HRP, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-68347H
This item is not returnable.
View return policy
Description
IgA Polyclonal antibody specifically detects IgA in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry
Specifications
| IgA | |
| Polyclonal | |
| PBS | |
| Sheep | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Bovine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry | |
| HRP | |
| Immunoglobulin A | |
| Purified bovine IgA | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| 3493 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction