Learn More
Sheep anti-Bovine IgA Heavy Chain Secondary Antibody, HRP, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-46602H
211.65 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Description
Specifically detects IgA in Bovine samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blotting
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry.
Specifications
| IgA Heavy Chain | |
| Polyclonal | |
| HRP | |
| Phosphate Buffered Saline (PBS) containing 0.2% BSA. with 0.05% Pro-Clean 400 | |
| IGHA1 | |
| Bovine IgA | |
| 1 mL | |
| Secondary | |
| By immunoelectrophoresis and ELISA this antibody reacts specifically with bovine IgA. This antibody may cross react with IgA from other species. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:20000, ELISA 1:10000 - 1:100000, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry 1:200 - 1:2000 | |
| Immunoglobulin A | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3493 | |
| Bovine | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only