missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78136
This item is not returnable.
View return policy
Description
SHD Polyclonal specifically detects SHD in Drosophila samples. It is validated for Western Blot, ELISA.
Specifications
| SHD | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:10000 | |
| Q9VUF8 | |
| SHD | |
| Synthetic peptide taken form the Spook protein form within the N-terminal region (amino acid 51-60) the peptide was post-synthetically modified and conjugated to carrier protein to achieve desired antigenicity. The Shade peptides sequences are conserved various other specieis of fruit flys. | |
| 0.1 mg | |
| Primary | |
| The species cross reactivity for this antibody is not examined fully. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| SH2 domain-containing adapter protein D, Src homology 2 domain containing transforming protein D, src homology 2 domain-containing transforming protein D | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 56961 | |
| Drosophila | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction