missing translation for 'onlineSavingsMsg'
Learn More

SHD Antibody, Novus Biologicals™

Product Code. 18226519 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
Unit Size:
0.05mg
0.1mg
25µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18226519 0.025 mg 25µg
18376961 0.05 mg 0.05mg
18287755 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18226519 Supplier Novus Biologicals Supplier No. NBP1781360.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SHD Polyclonal specifically detects SHD in Drosophila samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHD
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Accession No. Q9VUF8
Gene Alias SH2 domain-containing adapter protein D, Src homology 2 domain containing transforming protein D, src homology 2 domain-containing transforming protein D
Gene Symbols SHD
Host Species Rabbit
Immunogen Synthetic peptide taken form the Spook protein form within the N-terminal region (amino acid 51-60) the peptide was post-synthetically modified and conjugated to carrier protein to achieve desired antigenicity. The Shade peptides sequences are conserved various other specieis of fruit flys.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56961
Test Specificity The species cross reactivity for this antibody is not examined fully.
Target Species Drosophila
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.