missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£196.00 - £558.00
Specifications
| Antigen | SHD |
|---|---|
| Dilution | Western Blot 1:500, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18381020
|
Novus Biologicals
NBP1-78137-0.025MG |
0.025 mg |
£196.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18250529
|
Novus Biologicals
NBP1-78137-0.05MG |
0.05 mg |
£341.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18219065
|
Novus Biologicals
NBP1-78137 |
0.1 mg |
£558.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHD Polyclonal specifically detects SHD in Drosophila samples. It is validated for Western Blot, ELISA.Specifications
| SHD | |
| Western Blot, ELISA | |
| Unconjugated | |
| Drosophila | |
| SH2 domain-containing adapter protein D, Src homology 2 domain containing transforming protein D, src homology 2 domain-containing transforming protein D | |
| SHD | |
| IgG |
| Western Blot 1:500, ELISA 1:10000 | |
| Polyclonal | |
| Rabbit | |
| Q9VUF8 | |
| 56961 | |
| Synthetic peptide taken form the Spook protein form within the mid-region peptide (from with in aa 175-225), the peptides were post-synthetically modified and conjugated to carrier protein to achieve desired antigenicity. The Shade peptides sequences are conserved various other specieis of fruit flys. | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title