missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522161
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, Molt-4. Predicted reactivity: Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Activates both Ras-dependent and Ras-independent migratory pathways in melanomas. Contributes to the early phases of agrin-induced tyrosine phosphorylation of CHRNB1.
Specifications
| SHC4 | |
| Polyclonal | |
| Unconjugated | |
| SHC4 | |
| 6230417E10Rik; 9930029B02Rik; Gm685; hShcD; mShcD; novel phosphotyrosine interaction domain (PTB/PID) and SH2 domains containing protein; rai-like protein; rai-like protein RaLP; RaLP; RGD1309182; SH2 domain protein C4; SHC (Src homology 2 domain containing) family member 4; SHC (Src homology 2 domain containing) family, member 4; SHC adaptor protein 4; Shc4; Shcd; SHC-transforming protein 4; SHC-transforming protein D; src homology 2 domain-containing-transforming protein C4; UNQ6438/PRO21364 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 399694 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.54 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q6S5L8 | |
| SHC4 | |
| Recombinant protein encompassing a sequence within the center region of human SHC4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction