missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565099
This item is not returnable.
View return policy
Description
Immunogen sequence: HPYYNSIPSKM PPPGGFLDTR LKPRPHAPDT AQFAGKEQTY YQGRHLGDTF GEDWQQTPLR QGSSDIYSTP EGKLHVAPTG EAPTYVNTQQ IPP Highest antigen sequence identity to the following orthologs - mouse 81%, rat 81%.
Signaling adapter that couples activated growth factor receptors to signaling pathway in neurons. Involved in the signal transduction pathways of neurotrophin-activated Trk receptors in cortical neurons.
Specifications
| SHC3 | |
| Polyclonal | |
| Unconjugated | |
| SHC3 | |
| neuronal Shc; Nshc; N-Shc; Protein Rai; RAI; SH2 domain protein C3; SHC (Src homology 2 domain containing) transforming protein 3; SHC adaptor protein 3; SHC3; Shc3 p51; SHCC; SHC-transforming protein 3; SHC-transforming protein C; src homology 2 domain-containing transforming protein C3; src homology 2 domain-containing-transforming protein C3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53358 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92529 | |
| SHC3 | |
| Recombinant Human SHC3. Recombinant protein control fragment (Product #RP-96520). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction