missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | SHC2 |
|---|---|
| Dilution | Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670752
|
Novus Biologicals
NBP2-98061-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627624
|
Novus Biologicals
NBP2-98061-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHC2 Polyclonal antibody specifically detects SHC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SHC2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.0) | |
| 25759 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| neuronal Shc adaptor homolog, Protein Sck, SCKSH2 domain protein C2, SHC (Src homology 2 domain containing) transforming protein 2, SHCBSHC-transforming protein B, SHC-transforming protein 2, SLI, Src homology 2 domain-containing-transforming protein C2 | |
| Produced in rabbits immunized with E. coli-derived Human SHC2 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title