missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-18908
This item is not returnable.
View return policy
Description
SHC1 Polyclonal specifically detects SHC1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| SHC1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| FLJ26504, p66, p66SHC, SH2 domain protein C1, SHC, SHC (Src homology 2 domain containing) transforming protein 1, SHC (Src homology 2 domain-containing) transforming protein 1, SHCA, SHC-transforming protein 1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 1:10-1:500, Western Blot (Negative) | |
| NP_892113.4 | |
| SHC1 | |
| The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human SHC (Src homology 2 domain containing) transforming protein 1 using the numbering given in entry NP_892113.4 (GeneID 6464). | |
| 100 μL | |
| Cell Cycle and Replication, Signal Transduction | |
| 6464 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction