missing translation for 'onlineSavingsMsg'
Learn More

SHC1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18432804 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432804 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432804 Supplier Novus Biologicals Supplier No. NBP118908AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SHC1 Polyclonal antibody specifically detects SHC1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHC1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ26504, p66, p66SHC, SH2 domain protein C1, SHC, SHC (Src homology 2 domain containing) transforming protein 1, SHC (Src homology 2 domain-containing) transforming protein 1, SHCA, SHC-transforming protein 1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human SHC (Src homology 2 domain containing) transforming protein 1 using the numbering given in entry NP_892113.4 (GeneID 6464).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6464
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.