missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHBG Monoclonal Antibody (H3), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551095
This item is not returnable.
View return policy
Description
SHBG is a steroid binding protein that was first described as a plasma protein secreted by the liver and is thought to participate in the regulation of steroid responses. SHBG transports androgens and estrogens in the blood, binding each steroid molecule as a dimer formed from identical or nearly identical monomers. Low plasma SHBG levels are associated with obesity, abdominal adiposity, metabolic syndrome, and predict the development of type 2 diabetes. Polymorphisms in this gene have been associated with polycystic ovary syndrome and type 2 diabetes mellitus.
Specifications
| SHBG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P04278 | |
| SHBG | |
| Recombinant Human SHBG protein, Gly224-His388 (Accession #P04278). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| H3 | |
| Unconjugated | |
| SHBG | |
| ABP; Abpa; androgen binding protein; Androgen binding protein (ABP); SBP; sex hormone binding globulin; Sex hormone binding globulin or androgen-binding protein (other gene product from ABP gene); sex hormone-binding globulin; Sex hormone-binding globulin (SHBG); Sex steroid binding protein (SBP); Sex steroid-binding protein; Shbg; TeBG; testis-specific androgen-binding protein; Testosterone binding beta globulin; testosterone-binding beta-globulin; testosterone-estradiol-binding globulin; Testosterone-Estradiol-Binding Globulin (TEBG); Testosterone-estrogen-binding globulin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6462 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction