missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549181
This item is not returnable.
View return policy
Description
SHB Polyclonal Antibody for Western Blot
The SH2 (Src Homology 2) domain is a structurally conserved motif that contains two alpha helices and seven beta strands and is found in a variety of proteins that are involved in signal transduction throughout the cell. Specifically, the SH2 domain targets SH2 domain-containing proteins to tyrosinephosphorylated sites, an event that can trigger a protein-protein interaction cascade which may ultimately effect gene expression and cellular function. Shb (SH2 domain-containing adapter protein b), Shd (SH2 domain-containing adapter protein d), She (SH2 domain-containing adapter protein e) and Shf(SH2 domain-containing adapter protein f) are SH2 domain-containing proteins that play various roles throughout the cell.
Specifications
| SHB | |
| Polyclonal | |
| Unconjugated | |
| SHB | |
| bA3J10.2; BC028832; SH2 domain containing adaptor protein B; SH2 domain-containing adapter protein B; Shb; SHB (Src homology 2 domain containing) adaptor protein B; SHB adaptor protein (a Src homology 2 protein); Src homology 2 domain containing adaptor protein B; src homology 2 domain-containing transforming protein B | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 230126 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6PD21 | |
| SHB | |
| KLH conjugated synthetic peptide between 68-95 amino acids from the C-terminal region of mouse Shb. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction