missing translation for 'onlineSavingsMsg'
Learn More

SHB Antibody [PE], Novus Biologicals™

Product Code. 30734169 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734169 Supplier Novus Biologicals Supplier No. AF7036P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

SHB Polyclonal antibody specifically detects SHB in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHB
Applications Immunohistochemistry
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias bA3J10.2, RP11-3J10.8, SH2 domain-containing adapter protein B, SHB (Src homology 2 domain containing) adaptor protein B, SHB adaptor protein (a Src homology 2 protein), Src homology 2 domain containing adaptor protein B
Host Species Goat
Immunogen E. coli-derived recombinant human SHB, Ala2-Phe128, Accession # Q15464
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6461
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.