missing translation for 'onlineSavingsMsg'
Learn More

Sharpsafe Sharps container 4120A rectangular 24.0L

Brand:  Sharpsafe 41202430

Sharps container 4120A rectangular 24.0L

Code : 02

Additional Details : Weight : 1.00000kg

Product Code. 12963535

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.