Learn More
Sharpsafe™ Sharps Bin, Cytotoxic and Cytostatic Sharps
Description
- Wide aperture facilitates easy disposal.
- Translucent lid, clear internal flap and visible fill line help ensure that the bin is not overfilled.
- In-mold label that cannot be removed.
- Simple four-click assembly.
- Large headroom.
- Inkjet-printed fill line and waste segregation message.
- Temporary and final closure feature
Specifications
Specifications
| Capacity (Metric) | 2 L |
| Certifications/Compliance | UN 3291, BS 7320 and French AFNOR NX30-500 Standards |
| Closure Color | Purple |
| Quantity | 50/Cs. |
| For Use With (Application) | Cytotoxic and cytostatic waste collection and incineration. |