missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK3 Antibody (S367-51), Novus Biologicals™
Mouse Monoclonal Antibody
£176.00 - £427.00
Specifications
| Antigen | SHANK3 |
|---|---|
| Clone | S367-51 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692558
|
Novus Biologicals
NBP2-42189 |
0.1 mg |
£427.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629306
|
Novus Biologicals
NBP2-42189-0.025mg |
0.025 mg |
£176.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHANK3 Monoclonal antibody specifically detects SHANK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SHANK3 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| PBS (pH 7.4), 50% Glycerol | |
| DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2 | |
| Fusion protein amino acids 538-626 (SH3 domain) of rat SHANK3. Mouse: 100% identity (89/89 amino acids identical). Human: 97% identity (87/89 amino acids identical). ∽70% identity with SHANK1 and SHANK2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S367-51 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q9JLU4 | |
| 85358 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title