missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK2 Antibody (N23b/6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12914-0.025mg
This item is not returnable.
View return policy
Description
SHANK2 Monoclonal specifically detects SHANK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications
| SHANK2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-10 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 0.1-1.0 μg/mL | |
| SHANK2 | |
| Fusion protein amino acids 84-309 (SH3/PDZ domains) of rat Shank2 (SH3 and multiple ankyrin repeat domains protein 2) | |
| Protein G purified | |
| RUO | |
| 22941 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| S23b-6 | |
| Unconjugated | |
| AUTS17, cortactin binding protein 1, cortactin SH3 domain-binding protein, Cortactin-binding protein 1, CortBP1, CORTBP1CTTNBP1, GKAP/SAPAP interacting protein, KIAA1022, proline-rich synapse associated protein 1, Proline-rich synapse-associated protein 1, PROSAP1, SH3 and multiple ankyrin repeat domains 2, SH3 and multiple ankyrin repeat domains protein 2, SHANK, Shank2, SPANK-3 | |
| Mouse | |
| 159 kDa | |
| 0.025 mg | |
| Primary | |
| Detects approx 160kDa. No cross-reactivity against Shank1 or Shank3. Weak mouse detection. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction