missing translation for 'onlineSavingsMsg'
Learn More

SHANK2 Antibody [HRP], Novus Biologicals™

Product Code. 30735952 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735952 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735952 Supplier Novus Biologicals Supplier No. AF7035H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

SHANK2 Polyclonal antibody specifically detects SHANK2 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHANK2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias AUTS17, cortactin binding protein 1, cortactin SH3 domain-binding protein, Cortactin-binding protein 1, CortBP1, CORTBP1CTTNBP1, GKAP/SAPAP interacting protein, KIAA1022, proline-rich synapse associated protein 1, Proline-rich synapse-associated protein 1, PROSAP1, SH3 and multiple ankyrin repeat domains 2, SH3 and multiple ankyrin repeat domains protein 2, SHANK, Shank2, SPANK-3
Host Species Sheep
Immunogen E. coli-derived recombinant human SHANK2, Lys849-Thr1029, Accession # Q9UPX8
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 22941
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.