Learn More
Epredia™ Embedding Tissue Cassettes
Hold and identify tissue samples in processing, embedding, and sectioning procedures with Epredia™ Embedding Tissue Cassettes.
Brand: Epredia™ B1003500BL
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Designed to accept metal lids
- Designed with round hole pores
- Supplied in boxes of 500 cassettes
Specifications
Round Holes | |
Blue | |
Tissue Cassette |