missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Straight Blunt/Sharp Dissecting Scissors, premium, 6.5 in. (16.5cm)
Epredia™ Shandon Straight Blunt/Sharp Dissecting Scissors are available in Premium and Standard grades.
Brand: Epredia™ 28253
This item is not returnable.
View return policy
Description
Consistent Quality
- Properly hardened, tempered and passivated
- Electronically polished and cleaned for a high-quality finish
- Patterns, dimensions and finish held to exacting standards
- Guaranteed to be free from defects in material and workmanship
Specifications
Dissecting Scissors | |
Standard | |
16.5 cm |
Straight | |
6.5 in. | |
Blunt/Sharp |