Learn More
Epredia™ Shandon™ Polysine Slides, White
Product Code. 12647976 Shop All Epredia Products
Click to view available options
Description:
Blue; 144/Pack
White; 144/Pack
White; 1440/Case
Unit Size:
Pack of 144
Pack of 1440
Product Code. 12647976
Brand: Epredia™ 6776216
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Achieve superior cell and tissue adhesion with Epredia™ Shandon™ Polysine Slides.
  • Polysine places an advanced permanent adhesive on the microscope slide
  • Built-in chemicals electrostatically and chemically attract tissues and cells
TRUSTED_SUSTAINABILITY
Specifications
Material Polysine
Type Microscope Slide
Color Blue
Description Blue; 144/Pack