Learn More
Epredia™ Shandon™ Polysine Slides, White
Shop All Epredia Products
Click to view available options
Description:
Blue; 144/Pack
White; 144/Pack
White; 1440/Case
Unit Size:
Pack of 144
Pack of 1440
Description
- Polysine places an advanced permanent adhesive on the microscope slide
- Built-in chemicals electrostatically and chemically attract tissues and cells
Specifications
Specifications
| Material | Polysine |
| Type | Microscope Slide |
| Color | White |
| Description | White; 144/Pack |