missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ OG-6 Stain

Product Code. 10239788 Shop All Epredia Products
Click to view available options
Quantity:
1L
4L
Unit Size:
4 L
Pack of 2
This item is not returnable. View return policy

Product Code. 10239788

Brand: Epredia™ 6766006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain maximum performance and maximum value with Epredia™ Shandon™ OG-6 Stain, a biological stain commission-certified dye.

TRUSTED_SUSTAINABILITY

Specifications

Type OG-6 Papanicolaou Stain
Quantity 4L
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : methyl alcohol, ethylene glycol, orange-g certified, phosphotungstic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.