missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Gill™ Hematoxylin

Product Code. 12677926 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4L
2 x 1L
Type:
Shandon Gill 1 Hematoxylin
Shandon Gill 2 Hematoxylin
Shandon Gill 3 Hematoxylin
Unit Size:
4 L
Each
Pack of 2
Product Code. Quantity Type unitSize
12677926 2 x 1L Shandon Gill 3 Hematoxylin Pack of 2
12657926 2 x 1L Shandon Gill 1 Hematoxylin Pack of 2
10832130 4L Shandon Gill 1 Hematoxylin 4 L
10096648 4L Shandon Gill 2 Hematoxylin 4 L
10188158 4L Shandon Gill 3 Hematoxylin Each
5 options
This item is not returnable. View return policy

Product Code. 12677926

Brand: Epredia™ 6765009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Achieve fast and convenient results, while maintaining complete control over stain intensity with Epredia™ Shandon™ Gill™ Hematoxylin, which has mucoid staining.

Gill 3: Blue-black nuclei

TRUSTED_SUSTAINABILITY

Specifications

Type Shandon Gill 3 Hematoxylin
Quantity 2 x 1L
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : ethylene glycol, acetic acid, aluminiumsulphate, sodium iodate, hematoxylin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.