missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Frozen Section Staining Kit

Product Code. 12667716 Shop All Epredia Products
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12667716

Brand: Epredia™ 145

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Process a limited number of slides in an ideal environment with the Epredia™ Shandon™ Frozen Section Staining Kit.

  • Sturdy cover with piano hinge seals beakers to minimize evaporation
  • Rubber feet on rack protect table surface
  • Glass beakers have a capacity of 200mL (6.75 oz.) and are 10cm (4 in.) tall
  • Slide holder will hold ten 76 x 26mm (3 x 1 in.) slides
  • Stainless-steel support rack measures 3.5 x 4.5 x 17.25 in. when rack is closed

TRUSTED_SUSTAINABILITY

Specifications

Type Shandon Frozen Section Staining Kit
Includes Kit includes 12 beakers, a stainless-steel slide holder, and a heavy-gauge stainless-steel support rack with six round openings to hold beakers.
Quantity 1 Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.