missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Formal-Fixx™ 10% Neutral Buffered Formalin
Produce consistent results and extend life of rotary valve in tissue processors with Epredia™ Formal-Fixx™ 10% Neutral Buffered Foramalin.
Brand: Epredia™ 9990916
Code : NEW
Additional Details : Weight : 10.43273kg
Description
- Phosphate-free
- Use for storage of all tissue types
- Stores specimens without excessive shrinking or hardening
- Available premixed, ready-to-use, or in economical concentrate
- Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
- Pre-filled, leak-resistant cups are available
Specifications
90 mL Specimen Container filled to 45 mL | |
125/Cs. | |
Formalin | |
90 mL |