Learn More
Epredia™ Formal-Fixx™ 10% Neutral Buffered Formalin
Produce consistent results and extend life of rotary valve in tissue processors with Epredia™ Formal-Fixx™ 10% Neutral Buffered Foramalin.
Brand: Epredia™ 9990914
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Phosphate-free
- Use for storage of all tissue types
- Stores specimens without excessive shrinking or hardening
- Available premixed, ready-to-use, or in economical concentrate
- Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
- Pre-filled, leak-resistant cups are available
Specifications
125/Cs. | |
Formalin | |
60 mL specimen container filled to 30mL |
Safety and Handling
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- H370-Causes damage to organs.
Precautionary Statement
- P362-Take off contaminated clothing.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formaldehyde, methylalcohol, maleic acid, sodium hydroxide