Learn More
Epredia™ EZ Double Cytofunnel™
Product Code. p-4524835 Shop All Epredia Products
Click to view available options
Quantity:
200/Cs.
40 Pk.
Unit Size:
Pack of 200
Pack of 40
Product Code. 11911788
Brand: Epredia™ A78710005
Only null left
Add to basket
Edge
Add to basket
Double the number of sample presentations per run with the Epredia™ EZ Double Cytofunnel™.
  • Disposable sample chambers with white filter cards and caps
  • Funnel markings corresponding to slide markings for simple identification
  • Pre-attached filter card
TRUSTED_SUSTAINABILITY
Specifications
Color White
Disposable Single-use
Quantity 40 Pk.
Type EZ Double Cytofunnel™, With Filter Cards and Caps
Volume (Metric) 0.05 to 0.25 mL