Learn More
Epredia™ Shandon™ Eosin-Y Stain
Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
Brand: Epredia™ 6766010
176.25 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Code : LE
Additional Details : Weight : 3.84300kg
Description
- Ready-to-use
- Produces consistent results
- Stable and precipitate-free; no filtering
- Compatible with all alum hematoxylin stains
- Available in alcoholic or aqueous formulations
Specifications
Aqueous Eosin Y Stain | |
Each |
Safety and Handling
- Warning
Hazard Category
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H371-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P313-Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formaldehyde, aceticaid, sodium acetate, eosin-Y dye