Learn More
Epredia™ Shandon™ Embedding Cassettes II in Tube Packs, aqua
Hold tissue specimens during processing and embedding with Epredia™ Shandon™ Embedding Cassettes II in Tube Packs.
Brand: Epredia™ B1000729GY
Code : 04
Additional Details : Weight : 2.50000kg
Description
- Acetal polymer cassettes with writing face at 45° angle
- Slotted pore style
- Locking lid attached by two hinges
Ordering Information:
- Tube packs are supplied in 10 tubes of 75 cassettes each
Specifications
Slotted Pore | |
Gray | |
Biopsy Cassette |
45° | |
Acetal Polymer |