Learn More
Epredia™ Shandon™ Embedding Cassettes II in Tube Packs, aqua
Product Code. 12903755 Shop All Thermo Scientific Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
Product Code. 12903755
Brand: Epredia™ B1000729AQ
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Hold tissue specimens during processing and embedding with Epredia™ Shandon™ Embedding Cassettes II in Tube Packs.
  • Acetal polymer cassettes with writing face at 45° angle
  • Slotted pore style
  • Locking lid attached by two hinges
Ordering Information:
  • Tube packs are supplied in 10 tubes of 75 cassettes each
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slotted Pore
Writing Face Angle 45°
Color Aqua
Material Acetal Polymer
Product Type Biopsy Cassette