Learn More
Epredia™ Shandon™ CytoRich™ Red Collection Fluid
Allow for more accurate evaluation of cytological features with Epredia™ Shandon™ CytoRich™ Red Collection Fluid, which lyses RBCs and solubilizes proteins.
Brand:  Epredia™ B9990803
Code : 50
Additional Details : Weight : 7.50000kg Transport : UN number : 1987 Chem class : 3 Pack group : III
 View more versions of this product
Product Code. 12638346
  • £317.00 / Pack of 125
Estimated Shipment: 09-08-2024
to see stock.
Description
Description
  • Pre-filled containers available
  • Ready-to-use, alcohol-based fluid
  • Compatible with immunohistochemistry
Specifications
Specifications
Collection Fluid
120 mL